Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(4-Methoxy-2-methylphenyl)-3-(3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)urea 97%

Supplier: AMBEED, INC
Warning

A723107-1G A723107-250MG A723107-5G
77145-986EA 223.51 USD InStock
77145-986 77145-988 77145-990
1-(4-Methoxy-2-methylphenyl)-3-(3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)urea 97%
1-(4-Methoxy-2-methylphenyl)-3-(3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)urea
Precaución: for Research Use Only


Formula: C₂₁H₂₇BN₂O₄
MDL Number: MFCD20231480
CAS Number: 2096997-05-4
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty