Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,3-Dibromo-5-chloro-2-nitrobenzene 97%

Supplier: AMBEED, INC
Warning

A724220-100MG A724220-1G A724220-250MG
77145-998EA 17.52 USD InStock
77145-998 77146-000 77146-002
1,3-Dibromo-5-chloro-2-nitrobenzene 97%
1,3-Dibromo-5-chloro-2-nitrobenzene
Precaución: for Research Use Only


Formula: C₆H₂Br₂ClNO₂
MDL Number: MFCD24539371
CAS Number: 1824388-07-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty