Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3-Difluoro-4-(trifluoromethyl)benzeneboronic acid 96%

Supplier: AMBEED, INC
Danger

A726844-1G A726844-250MG A726844-5G
77146-016EA 126.69 USD InStock
77146-016 77146-018 77146-020
2,3-Difluoro-4-(trifluoromethyl)benzeneboronic acid 96%
2,3-Difluoro-4-(trifluoromethyl)benzeneboronic acid
Precaución: for Research Use Only


Formula: C₇H₄BF₅O₂
MW: 225.908 g/mol
MDL Number: MFCD14892600
CAS Number: 2096332-61-3
UN: 2811
ADR: 6.1 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty