Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(((Benzyloxy)carbonyl)amino)-5-methylhexanoic acid 95%

Supplier: AMBEED, INC
Warning

A731836-100MG A731836-1G A731836-250MG
77146-068EA 24.73 USD InStock
77146-068 77146-070 77146-072
3-(((Benzyloxy)carbonyl)amino)-5-methylhexanoic acid 95%
3-(((Benzyloxy)carbonyl)amino)-5-methylhexanoic acid
Precaución: for Research Use Only


Formula: C₁₅H₂₁NO₄
MDL Number: MFCD19382359
CAS Number: 1311254-63-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty