Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ferene disodium salt for spectrophotometric determination of Fe Emax 593nm Fe2+/Ferene>34000

Supplier: AMBEED, INC
Warning

A738859-1G A738859-250MG A738859-25G A738859-5G
77146-106EA 35.02 USD InStock
77146-106 77146-108 77146-110 77146-112
Ferene disodium salt for spectrophotometric determination of Fe Emax 593nm Fe2+/Ferene>34000
Ferene disodium salt
Precaución: for Research Use Only


Formula: C₁₆H₈N₄Na₂O₈S₂
MW: 494.37 g/mol
MDL Number: MFCD00040642
CAS Number: 79551-14-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty