Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

L-α-Hydroxyglutaric Acid Disodium Salt 95%

Supplier: AMBEED, INC
Warning

A754859-1G A754859-1MG A754859-5G
77146-266EA 29.87 USD InStock
77146-266 77146-268 77146-270
L-α-Hydroxyglutaric Acid Disodium Salt 95%
L-α-Hydroxyglutaric Acid Disodium Salt
Precaución: for Research Use Only


Formula: C₅H₆Na₂O₅
MW: 192.08 g/mol
MDL Number: MFCD00021700
CAS Number: 63512-50-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty