Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-2-ethynylpyridine 97%

Supplier: AMBEED, INC
Warning

A758874-100MG A758874-1G A758874-250MG
77146-290EA 35.02 USD InStock
77146-290 77146-292 77146-294
3-Bromo-2-ethynylpyridine 97%
3-Bromo-2-ethynylpyridine
Precaución: for Research Use Only


Formula: C₇H₄BrN
MDL Number: MFCD18256373
CAS Number: 96439-99-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty