Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-2-fluoro-3-hydroxypyridine 98%

Supplier: AMBEED, INC
Warning

A774796-10G A774796-1G A774796-250MG A774796-25G A774796-5G
77146-390EA 86.52 USD InStock
77146-390 77146-392 77146-394 77146-396 77146-398
5-Bromo-2-fluoro-3-hydroxypyridine 98%
5-Bromo-2-fluoro-3-hydroxypyridine
Precaución: for Research Use Only


Formula: C₅H₃BrFNO
MW: 191.986 g/mol
MDL Number: MFCD11040265
CAS Number: 1012084-53-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty