Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 2-bromo-4-cyanobenzoate 95%

Supplier: AMBEED, INC
Warning

A775874-1G A775874-250MG A775874-5G
77146-406EA 112.27 USD InStock
77146-406 77146-408 77146-410
Methyl 2-bromo-4-cyanobenzoate 95%
Methyl 2-bromo-4-cyanobenzoate
Precaución: for Research Use Only


Formula: C₉H₆BrNO₂
MDL Number: MFCD18398738
CAS Number: 98592-04-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty