Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-2-Ethyl-piperazine 98%

Supplier: AMBEED, INC
Danger

A779346-100MG A779346-1G A779346-250MG A779346-5G
77146-432EA 6.18 USD InStock
77146-432 77146-434 77146-436 77146-438
(S)-2-Ethyl-piperazine 98%
(S)-2-Ethyl-piperazine
Precaución: for Research Use Only


Formula: C₆H₁₄N₂
MW: 114.189 g/mol
MDL Number: MFCD07373041
CAS Number: 207284-20-6
UN: 3259
ADR: 8 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty