Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Fluoro-3-(trifluoromethyl)phenol 98%

Supplier: AMBEED, INC
Danger

A790981-10G A790981-1G A790981-25G A790981-5G
77146-510EA 17.51 USD InStock
77146-510 77146-512 77146-514 77146-516
4-Fluoro-3-(trifluoromethyl)phenol 98%
4-Fluoro-3-(trifluoromethyl)phenol
Precaución: for Research Use Only


Formula: C₇H₄F₄O
MW: 180.1 g/mol
Boiling Pt: 84
MDL Number: MFCD00043875
CAS Number: 61721-07-1
UN: 3265
ADR: 8 Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty