Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-(4-Cyano-3-(trifluoromethyl)phenyl)-3-((2-fluorophenyl)sulfonyl)-2-hydroxy-2-methylpropanamide 97%

Supplier: AMBEED, INC
Warning

A796689-100MG A796689-250MG A796689-1G
77146-550EA 93.73 USD InStock
77146-550 77146-552 77308-348
N-(4-Cyano-3-(trifluoromethyl)phenyl)-3-((2-fluorophenyl)sulfonyl)-2-hydroxy-2-methylpropanamide 97%
N-(4-Cyano-3-(trifluoromethyl)phenyl)-3-((2-fluorophenyl)sulfonyl)-2-hydroxy-2-methylpropanamide
Precaución: for Research Use Only


Formula: C₁₈H₁₄F₄N₂O₄S
MDL Number: MFCD28898396
CAS Number: 1159977-36-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty