Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3-Difluorobenzylamine 98%

Supplier: AMBEED, INC
Danger

A803751-1G A803751-250MG A803751-5G
77146-600EA 46.35 USD InStock
77146-600 77146-602 77146-604
2,3-Difluorobenzylamine 98%
2,3-Difluorobenzylamine
Precaución: for Research Use Only


Formula: C₇H₇F₂N
MW: 143.14 g/mol
Boiling Pt: 174
Melting Pt: 31
MDL Number: MFCD00070793
CAS Number: 72235-51-9
UN: 2735
ADR: 8 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty