Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-4-fluoro-N-methyl-6-nitroaniline 93%

Supplier: AMBEED, INC
Warning

A809079-1G A809079-250MG
77146-648EA 40.32 USD InStock
77146-648 77146-650 77146-652
2-Bromo-4-fluoro-N-methyl-6-nitroaniline 93%
2-Bromo-4-fluoro-N-methyl-6-nitroaniline
Precaución: for Research Use Only


Formula: C₇H₆BrFN₂O₂
MW: 249.03 g/mol
MDL Number: MFCD28962791
CAS Number: 1845693-81-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty