Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N,N,N',N'-Tetraethylmalonamide 98%

Supplier: AMBEED, INC
Warning

A811591-1G A811591-250MG A811591-25G A811591-5G
77146-658EA 29.86 USD InStock
77146-658 77146-660 77146-662 77146-664
N,N,N',N'-Tetraethylmalonamide 98%
N,N,N',N'-Tetraethylmalonamide
Precaución: for Research Use Only


Formula: C₁₁H₂₂N₂O₂
MDL Number: MFCD06797171
CAS Number: 33931-42-9
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty