Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Chloro-4-nitro-o-toluidine 97%

Supplier: AMBEED, INC
Warning

A821353-250MG A821353-25G A821353-5G A821353-100G
77146-702EA 6.18 USD InStock
77146-702 77146-704 77146-706 77747-573
6-Chloro-4-nitro-o-toluidine 97%
6-Chloro-4-nitro-o-toluidine
Precaución: for Research Use Only


Formula: C₇H₇ClN₂O₂
MW: 186.6
Melting Pt: 172.5
MDL Number: MFCD01313703
CAS Number: 69951-02-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty