Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Boc-D-Abu-OH DCHA 98%

Supplier: AMBEED, INC
Warning

A831164-100G A831164-25G A831164-5G
77146-788EA 52.53 USD InStock
77146-788 77146-790 77146-792
Boc-D-Abu-OH DCHA 98%
Boc-D-Abu-OH DCHA
Precaución: for Research Use Only


Formula: C₂₁H₄₀N₂O₄
MW: 384.56 g/mol
MDL Number: MFCD00235886
CAS Number: 27494-47-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty