Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,3-Difluoro-4-iodo-2-methylbenzene 97%

Supplier: AMBEED, INC
Warning

A840358-100MG A840358-1G A840358-250MG
77146-854EA 85.48 USD InStock
77146-854 77146-856 77146-858
1,3-Difluoro-4-iodo-2-methylbenzene 97%
1,3-Difluoro-4-iodo-2-methylbenzene
Precaución: for Research Use Only


Formula: C₇H₅F₂I
MDL Number: MFCD12922605
CAS Number: 1208077-89-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty