Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4,6-Tris(4-fluorophenyl)boroxin 98%

Supplier: AMBEED, INC
Warning

A846377-100G A846377-1G A846377-25G A846377-5G
77146-886EA 130.81 USD InStock
77146-886 77146-888 77146-890 77146-892
2,4,6-Tris(4-fluorophenyl)boroxin 98%
2,4,6-Tris(4-fluorophenyl)boroxin
Precaución: for Research Use Only


Formula: C₁₈H₁₂B₃F₃O₃
MDL Number: MFCD02093496
CAS Number: 448-59-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty