Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Brigatinib (AP26113) analog 98+%

Supplier: AMBEED, INC
Warning

A846689-10MG A846689-5MG
77146-894EA 7.21 USD InStock
77146-894 77146-896
Brigatinib (AP26113) analog 98+%
Brigatinib (AP26113) analog
Precaución: for Research Use Only


Formula: C₂₆H₃₄ClN₆O₂P
MW: 529.01 g/mol
MDL Number: MFCD23704187
CAS Number: 1197958-12-5
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty