Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-3-fluoro-1H-pyrazole 98%

Supplier: AMBEED, INC
Warning

A851291-1G A851291-250MG A851291-50MG A851291-100MG
77146-920EA 234.84 USD InStock
77146-920 77146-922 77146-924 77686-888
4-Bromo-3-fluoro-1H-pyrazole 98%
4-Bromo-3-fluoro-1H-pyrazole
Precaución: for Research Use Only


Formula: C₃H₂BrFN₂
MDL Number: MFCD28517446
CAS Number: 1346555-56-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty