Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dimethyl Dipropargylmalonate 97%

Supplier: AMBEED, INC
Warning

A851324-10G A851324-250MG A851324-25G A851324-5G A851324-1G
77146-926EA 334.75 USD InStock
77146-926 77146-928 77146-930 77146-932 77618-384
Dimethyl Dipropargylmalonate 97%
Dimethyl Dipropargylmalonate
Precaución: for Research Use Only


Formula: C₁₁H₁₂O₄
MW: 208.21 g/mol
MDL Number: MFCD00455571
CAS Number: 63104-44-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty