Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,5-Dichloro-4-fluoropyridine 97%

Supplier: AMBEED, INC
Warning

A872814-100MG A872814-1G A872814-250MG
77146-988EA 52.53 USD InStock
77146-988 77146-990 77146-992
2,5-Dichloro-4-fluoropyridine 97%
2,5-Dichloro-4-fluoropyridine
Precaución: For research use only.


Formula: C₅H₂Cl₂FN
MDL Number: MFCD13185533
CAS Number: 1214350-63-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty