Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(3,5-Dichlorophenyl)methanesulfonyl chloride 97%

Supplier: AMBEED, INC
Danger

A875687-100MG A875687-1G
77146-994EA 60.77 USD InStock
77146-994 77146-996
(3,5-Dichlorophenyl)methanesulfonyl chloride 97%
(3,5-Dichlorophenyl)methanesulphonyl chloride
Precaución: for Research Use Only


Formula: C₇H₅CI₃O₂S
MDL Number: MFCD01631928
CAS Number: 163295-70-3
UN: 3261
ADR: 8,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty