Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Aminomethyl)-3-bromobenzonitrile 98%

Supplier: AMBEED, INC
Danger

A899949-100MG A899949-250MG
77147-112EA 66.95 USD InStock
77147-112 77147-114
4-(Aminomethyl)-3-bromobenzonitrile 98%
4-(Aminomethyl)-3-bromobenzonitrile
Precaución: for Research Use Only


Formula: C₈H₇BrN₂
MW: 211.06 g/mol
MDL Number: MFCD18390755
CAS Number: 1261568-80-2
UN: 3259
ADR: 8 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty