Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(2-Chlorophenyl)acetohydrazide 98%

Supplier: AMBEED, INC
Warning

A904277-100MG A904277-1G A904277-250MG A904277-5G
77147-130EA 7.21 USD InStock
77147-130 77147-132 77147-134 77147-136
2-(2-Chlorophenyl)acetohydrazide 98%
2-(2-Chlorophenyl)acetohydrazide
Precaución: for Research Use Only


Formula: C₈H₉ClN₂O
MDL Number: MFCD00176721
CAS Number: 22631-60-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty