Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(4-Chlorophenyl)(dimethoxyphosphoryl)methyl dimethyl phosphate 98%

Supplier: AMBEED, INC
Warning

A913260-1MG A913260-5MG
77147-166EA 10.3 USD InStock
77147-166 77147-168
(4-Chlorophenyl)(dimethoxyphosphoryl)methyl dimethyl phosphate 98%
(4-Chlorophenyl)(dimethoxyphosphoryl)methyl dimethyl phosphate
Precaución: for Research Use Only


Formula: C₁₁H₁₇ClO₇P₂
MDL Number: MFCD00864813
CAS Number: 76541-72-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty