Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(N-Hydroxycarbamimidoyl)acetic acid ethyl ester 97%

Supplier: AMBEED, INC
Warning

A925523-250MG A925523-5G A925523-1G
77147-202EA 40.18 USD InStock
77147-202 77747-689 77747-688
(N-Hydroxycarbamimidoyl)acetic acid ethyl ester 97%
(N-Hydroxycarbamimidoyl)acetic acid ethyl ester
Precaución: for Research Use Only


Formula: C₅H₁₀N₂O₃
MDL Number: MFCD11227209
CAS Number: 89364-92-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty