Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2,4-Difluoro-5-hydroxyphenyl)boronic acid 97%

Supplier: AMBEED, INC
Warning

A946383-100MG A946383-1G A946383-250MG
77147-258EA 99.92 USD InStock
77147-258 77147-260 77147-262
(2,4-Difluoro-5-hydroxyphenyl)boronic acid 97%
(2,4-Difluoro-5-hydroxyphenyl)boronic acid
Precaución: for Research Use Only


Formula: C₆H₅BF₂O₃
MW: 173.90 g/mol
MDL Number: MFCD18783139
CAS Number: 2096330-91-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty