Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diethyl bromomalonate 98%

Supplier: AMBEED, INC
Danger

A971976-10G A971976-25G
77147-328EA 5.15 USD InStock
77147-328 77147-330
Diethyl bromomalonate 98%
Diethyl bromomalonate
Precaución: for Research Use Only


Formula: C₇H₁₁BrO₄
MDL Number: MFCD00009138
CAS Number: 685-87-0
UN: 3265
ADR: 8,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty