Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4,6-Tris(4-methoxyphenyl)pyrylium tetrafluoroborate 97%

Supplier: AMBEED, INC
Warning

A973789-1G A973789-250MG A973789-50MG
77147-332EA 486.16 USD InStock
77147-332 77147-334 77147-336
2,4,6-Tris(4-methoxyphenyl)pyrylium tetrafluoroborate 97%
2,4,6-Tris(4-methoxyphenyl)pyrylium tetrafluoroborate
Precaución: for Research Use Only


Formula: C₂₆H₂₃BF₄O₄
MDL Number: MFCD00160229
CAS Number: 580-34-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty