Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Chloro(dimethylsulfide)gold(I) 98%

Supplier: AMBEED, INC
Warning

A987858-1G A987858-250MG A987858-5G
77147-356EA 180.25 USD InStock
77147-356 77147-358 77147-360
Chloro(dimethylsulfide)gold(I) 98%
Chloro(dimethylsulfide)gold(I)
Precaución: for Research Use Only


Formula: C₂H₆AuClS
MDL Number: MFCD00192090
CAS Number: 29892-37-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty