Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl (3-hydroxy-4-methylphenyl)carbamate 97%

Supplier: AMBEED, INC
Warning

A101571-100MG A101571-10G A101571-1G A101571-250MG A101571-5G
77148-210EA 22 USD InStock
77148-210 77148-212 77148-214 77148-216 77148-218
tert-Butyl (3-hydroxy-4-methylphenyl)carbamate 97%
tert-Butyl (3-hydroxy-4-methylphenyl)carbamate
Precaución: for Research Use Only


Formula: C₁₂H₁₇NO₃
MW: 223.268 g/mol
MDL Number: MFCD20414278
CAS Number: 345893-26-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty