Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Enoxacin sesquihydrate 98%

Supplier: AMBEED, INC
Warning

A102347-100MG A102347-10G A102347-1G
77148-276EA 6 USD InStock
77148-276 77148-278 77148-280
Enoxacin sesquihydrate 98%
Enoxacin sesquihydrate
Precaución: for Research Use Only


Formula: C₃₀H₄₀F₂N₈O₉
MDL Number: MFCD01747755
CAS Number: 84294-96-2
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty