Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-((2,5-Dioxo-2,5-dihydro-1H-pyrrol-1-yl)methyl)-N-(prop-2-yn-1-yl)cyclohexanecarboxamide 97%

Supplier: AMBEED, INC
Warning

A103872-100MG A103872-1G A103872-250MG A103872-5G
77148-340EA 10 USD InStock
77148-340 77148-342 77148-344 77148-346
4-((2,5-Dioxo-2,5-dihydro-1H-pyrrol-1-yl)methyl)-N-(prop-2-yn-1-yl)cyclohexanecarboxamide 97%
4-((2,5-Dioxo-2,5-dihydro-1H-pyrrol-1-yl)methyl)-N-(prop-2-yn-1-yl)cyclohexanecarboxamide
Precaución: for Research Use Only


Formula: C₁₅H₁₈N₂O₃
MDL Number: MFCD27956997
CAS Number: 1036847-90-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty