Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Bromosporine 98%

Supplier: AMBEED, INC
Warning

A104099-10MG A104099-1MG A104099-5MG
77148-348EA 125 USD InStock
77148-348 77148-350 77148-352
Bromosporine 98%
Bromosporine
Precaución: for Research Use Only


Formula: C₁₇H₂₀N₆O₄S
MDL Number: MFCD26142669
CAS Number: 1619994-69-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty