Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Amino-3-(3-bromophenyl)propionic acid 97%

Supplier: AMBEED, INC
Warning

A109341-1G A109341-250MG A109341-25G A109341-5G
77148-634EA 7 USD InStock
77148-634 77148-636 77148-638 77148-640
3-Amino-3-(3-bromophenyl)propionic acid 97%
3-Amino-3-(3-bromophenyl)propionic acid
Precaución: for Research Use Only


Formula: C₉H₁₀BrNO₂
MW: 244.0852 g/mol
MDL Number: MFCD00995843
CAS Number: 117391-50-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty