Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Acetyl-S-trityl-L-cysteine 97%

Supplier: AMBEED, INC
Warning

A111243-10G A111243-1G A111243-250MG A111243-5G
77148-708EA 19 USD InStock
77148-708 77148-710 77148-712 77148-714
N-Acetyl-S-trityl-L-cysteine 97%
N-Acetyl-S-trityl-L-cysteine
Precaución: for Research Use Only


Formula: C₂₄H₂₃NO₃S
MDL Number: MFCD00236747
CAS Number: 27486-87-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty