Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(4-(Trifluoromethoxy)benzyl)hydrazine dihydrochloride 95%

Supplier: AMBEED, INC
Warning

A111509-100MG A111509-1G A111509-250MG
77148-724EA 28 USD InStock
77148-724 77148-726 77148-728
(4-(Trifluoromethoxy)benzyl)hydrazine dihydrochloride 95%
(4-(Trifluoromethoxy)benzyl)hydrazine dihydrochloride
Precaución: for Research Use Only


Formula: C₈H₁₁Cl₂F₃N₂O
MW: 279.08 g/mol
MDL Number: MFCD21642367
CAS Number: 1427416-69-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty