Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fmoc-Glu(OMe)-OH 97%

Supplier: AMBEED, INC
Warning

A116009-100G A116009-10G A116009-25G A116009-5G
77148-938EA 67 USD InStock
77148-938 77148-940 77148-942 77148-944
Fmoc-Glu(OMe)-OH 97%
Fmoc-Glu(OMe)-OH
Precaución: for Research Use Only


Formula: C₂₁H₂₁NO₆
MW: 383.4 g/mol
MDL Number: MFCD01076132
CAS Number: 145038-50-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty