Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Octyl benzoate 95%

Supplier: AMBEED, INC
Warning

A116245-1G A116245-250MG
77148-952EA 81 USD InStock
77148-952 77148-954
Octyl benzoate 95%
Octyl benzoate
Precaución: for Research Use Only


Formula: C₁₅H₂₂O₂
MDL Number: MFCD00995104
CAS Number: 94-50-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty