Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Amino-PEG3-acid 95%

Supplier: AMBEED, INC
Warning

A116310-100MG A116310-250MG A116310-1G A116310-5G A116310-25G
77148-956EA 12 USD InStock
77148-956 77148-958 77311-394 77311-396 77685-030
Amino-PEG3-acid 95%
Amino-PEG3-acid
Precaución: for Research Use Only


Formula: C₉H₁₉NO₅
MW: 221.251 g/mol
MDL Number: MFCD21607085
CAS Number: 784105-33-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty