Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

GFT505 95%

Supplier: AMBEED, INC
Warning

A118286-1MG A118286-5MG A118286-100MG
77149-016EA 14 USD InStock
77149-016 77149-018 77685-044
GFT505 95%
GFT505
Precaución: for Research Use Only


Formula: C₂₂H₂₄O₄S
MW: 384.39 g/mol
MDL Number: MFCD27987940
CAS Number: 923978-27-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty