Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chloro-6-fluoroisoquinoline 95%

Supplier: AMBEED, INC
Warning

A119900-100MG A119900-1G A119900-250MG
77149-150EA 18 USD InStock
77149-150 77149-152 77149-154
3-Chloro-6-fluoroisoquinoline 95%
3-Chloro-6-fluoroisoquinoline
Precaución: for Research Use Only


Formula: C₉H₅ClFN
MW: 181.59 g/mol
MDL Number: MFCD18837816
CAS Number: 1041423-28-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty