Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Dibromo-5-methoxyaniline 95%

Supplier: AMBEED, INC
Warning

A120653-10G A120653-1G A120653-250MG A120653-5G
77149-174EA 180 USD InStock
77149-174 77149-176 77149-178 77149-180
2,4-Dibromo-5-methoxyaniline 95%
2,4-Dibromo-5-methoxyaniline
Precaución: for Research Use Only


Formula: C₇H₇Br₂NO
MDL Number: MFCD08282833
CAS Number: 35736-52-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty