Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

C646 98%

Supplier: AMBEED, INC
Warning

A122218-10MG A122218-1MG A122218-50MG A122218-5MG
77149-248EA 23 USD InStock
77149-248 77149-250 77149-252 77149-254
C646 98%
C646
Precaución: for Research Use Only


Formula: C₂₄H₁₉N₃O₆
MW: 445.4 g/mol
MDL Number: MFCD01784780
CAS Number: 328968-36-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty