Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

O-Benzyl-D-Serine 97%

Supplier: AMBEED, INC
Warning

A124681-100G A124681-1G A124681-250MG A124681-5G
77149-368EA 221 USD InStock
77149-368 77149-370 77149-372 77149-374
O-Benzyl-D-Serine 97%
O-Benzyl-D-Serine
Precaución: For research use only.


Formula: C₁₀H₁₃NO₃
MDL Number: MFCD00065936
CAS Number: 10433-52-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty