Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

PF-06447475 98+%

Supplier: AMBEED, INC
Warning

A125109-10MG A125109-1MG A125109-25MG A125109-50MG A125109-5MG
77149-388EA 24 USD InStock
77149-388 77149-390 77149-392 77149-394 77149-396
PF-06447475 98+%
PF-06447475
Precaución: for Research Use Only


Formula: C₁₇H₁₅N₅O
MW: 305.33 g/mol
MDL Number: MFCD28347838
CAS Number: 1527473-33-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty