Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diphenyl(methyl)sulfonium tetrafluoroborate 95%

Supplier: AMBEED, INC
Danger

A128876-250MG A128876-25G A128876-5G
77149-514EA 7 USD InStock
77149-514 77149-516 77149-518
Diphenyl(methyl)sulfonium tetrafluoroborate 95%
Diphenyl(methyl)sulfonium tetrafluoroborate
Precaución: for Research Use Only


Formula: C₁₃H₁₃BF₄S
MDL Number: MFCD00051861
CAS Number: 10504-60-6
UN: 1759
ADR: 8,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty