Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(1S,2S,3R,4R)-3-((S)-1-Acetamido-2-ethylbutyl)-4-guanidino-2-hydroxycyclopentanecarboxylic acid trihydrate 98%

Supplier: AMBEED, INC
Warning

A130237-10MG A130237-1G A130237-250MG A130237-25G A130237-50MG A130237-5G
77149-562EA 6 USD InStock
77149-562 77149-564 77149-566 77149-568 77149-570 77149-572
(1S,2S,3R,4R)-3-((S)-1-Acetamido-2-ethylbutyl)-4-guanidino-2-hydroxycyclopentanecarboxylic acid trihydrate 98%
(1S,2S,3R,4R)-3-((S)-1-Acetamido-2-ethylbutyl)-4-guanidino-2-hydroxycyclopentanecarboxylic acid trihydrate
Precaución: for Research Use Only


Formula: C₁₅H₃₄N₄O₇
MDL Number: MFCD22417093
CAS Number: 1041434-82-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty